Raptor™ LC Cloumns – Selectivity Accelerated [DK]
Raptor™ ARC-18 LC Columns (USP L1)
- Ideal for high-throughput LC-MS/MS applications with minimal sample preparation.
- Well-balanced retention profile for better detection and integration of large, multiclass analyte lists.
- Sterically protected to endure low-pH mobile phases without sacrificing retention or peak quality.
- Part of Restek's Raptor™ LC column line featuring SPP core-shell silica.
Raptor™ Biphenyl LC Columns (USP L11)
- Ideal for bioanalytical testing applications like drug and metabolite analyses.
- Heightened selectivity and retention for compounds that are hard to resolve or elute early on C18 and other phenyl chemistries.
- Limits ionization suppression and allows simple, MS-friendly mobile phases.
- Restek's most popular LC stationary phase applied to Raptor™ SPP core-shell silica.
Please contact teamkemikalier@dk.vwr.com if you need furtther information or an offer on this new column species.