半胱胺盐酸盐, Pierce™

供应商: Thermo Fisher Scientific

Synonyms: 硫化乙醇胺2-巯基乙胺巯乙胺2-氨基乙硫醇β-巯基乙胺

PIER20408EA 0 CNY
PIER20408
半胱胺盐酸盐, Pierce™
半胱胺盐酸盐

Thermo Scientific Pierce 2-Mercaptoethylamine-HCl, commonly abbreviated 2-MEA, is a mild reductant that is often used to selectively reduce hinge-region disulfide bonds in antibody.


Caution: For Research Use Only. Not for use in diagnostic procedures.

分子式: C₂H₇NS·HCl
分子量: 113,61 g/mol
存储条件: Refrigerator
MDL编号: MFCD00012904
CAS号: 156-57-0
EINECS: 205-858-1

现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR