Eriochrome 铬氰蓝 RC | 铬花氰 R, 纯净

供应商: Thermo Scientific

ACRO215150050EA 0 CNY
ACRO215150050
Eriochrome 铬氰蓝 RC | 铬花氰 R, 纯净
Eriochrome 铬氰蓝 RC | 铬花氰 R
分子式: C₂₃H₁₅Na₃O₉S
存储条件: Ambient
MDL编号: MFCD00012411
CAS号: 3564-18-9
EINECS: 222-641-7

现在订购

产品技术规格

Appearance Red to dark brown Powder
Infrared spectrum Conforms
UV spectrum Complies

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR