2-硝基苯乙腈 ≥98.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAN0781-5GEA 0 CNY
TCIAN0781-5G
2-硝基苯乙腈 ≥98.0% (通过GC)
2-硝基苯乙腈
分子式: C₈H₆N₂O₂
分子量: 162,15 g/mol
沸点: 178°C
溶点: 81…83 °C
MDL编号: MFCD00007183
CAS号: 610-66-2
EINECS: 210-231-0
UN号: 3439
ADR: 6.1,III
https://cn.vwr.com/stibo/web/std.lang.all/99/85/28989985.jpg

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 82.0 to 86.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR