双硫腙 ≥85.0% (通过UV/Vis)
供应商: TCI EUROPE
|
Warning
|
Synonyms:
二苯基硫卡巴腙
TCIAD0949-25GEA
0
CNY
TCIAD0949-25G
双硫腙 ≥85.0% (通过UV/Vis)
双硫腙
分子式:
C₆H₅NHNHCSN=NC₆H₅ 分子量: 256,33 g/mol 溶点: 169°C |
MDL编号:
MFCD00003025 CAS号: 60-10-6 EINECS: 200-454-1 Merck Index: 14,03377 |
产品技术规格
Appearance | Red to Dark blue to Black powder to crystal |
Purity (UV-vis method) | min. 85.0 % |
abs-ratio (a/b) | min. 1.55 (E620/E450) |
Suitability test | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...