2-Nitrophenyl β-D-galactopyranoside, 超纯级

供应商: VWR Chemicals

Synonyms: ONPGo-Nitrophenyl β-D-galactopyranoside

VWRC0789-25GEA 2132 CNY
VWRC0789-25G
2-Nitrophenyl β-D-galactopyranoside, 超纯级
2-Nitrophenyl β-D-galactopyranoside

Chromogenic substrate for the detection of β-galactosidase. Commonly used in ELISA. Absorbs at 410 nm.

分子式: C₁₂H₁₅NO₈
分子量: 301,25 g/mol
溶点: 185…190 °C
存储条件: Freezer
MDL编号: MFCD00063255
CAS号: 369-07-3
EINECS: 206-716-1

现在订购

产品技术规格

Solubility (1%, Water) pass
Specific Rotation 185 - 190 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR