6-Azauracil 99%

供应商: Thermo Scientific
Danger

Synonyms: 6-Aza-2,4-dihydroxypyrimidine1,2,4-Triazine-3,5(2H,4H)-dione3,5-Dihydroxy-1,2,4-triazine

ACRO153970050EA 0 CNY
ACRO153970050
6-Azauracil 99%
6-Azauracil
分子式: C₃H₃N₃O₂
分子量: 113,08 g/mol
溶点: 276…280 °C
存储条件: Ambient
MDL编号: MFCD00006456
CAS号: 461-89-2
EINECS: 207-318-0
UN号: 2811
ADR: 6.1,II

现在订购

产品技术规格

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 276 to 280 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR