Spill containment tray, small, SP Bel-Art
供应商: Bel-Art
SP Bel-Art
BELAF189050001EA 0 CNY
BELAF189050001
Spill containment tray, small, SP Bel-Art
盘子/皿/盆 溢出托盘/锅
Made of PP, translucent.
  • Good chemical resistance
  • Durable
  • Steam autoclavable at 121 °C
Reduce injury and damage from accidental spills.