Biohazard waste bin, Touch Free™, SP Bel-Art
供应商: Bel-Art Products, a part of SP
SP Bel-Art Touch Free™
BELAF132020020EA 0 CNY
BELAF132020020
Biohazard waste bin, Touch Free™, SP Bel-Art
箱柜 垃圾箱
These Touch Free™ automatic waste bins protect from contact with hazardous waste. Detects the motion near the receptacle lid.
  • Approximately 4000 openings
  • On/off switch can deactivate the automatic open/close function
  • Lip of the lid designed to fit into a groove to reduce odor and airborne contamination
Delivery information: Supplied with a Biohazard label, plus three other labels for 'Recyclables', 'Trash' and a blank label. Batteries are not included.