Mercury collectors, SP Bel-Art
供应商: Bel-Art Products
BELAF362510000EA 0 CNY
BELAF362510000 BELAF362500000
Mercury collectors, SP Bel-Art
泄露控制装置
The mercury globules are absorbed by the foam pad.
  • Deals safely and effectively with mercury spillages
  • Safe storage in airtight container
  • Mesh inside squeezes the mercury out of the foam, so it is immediately ready for use again