Alcohol burners
供应商: DWK Life Sciences
WHEA237070EA 0 CNY
WHEA237070
Alcohol burners
加热器 酒精灯
Alcohol burner manufactured from soda-lime glass, designed for use with isopropyl or denatured ethyl alcohol.
  • Low-heat flame is well suited for microscopy purposes
  • With a ground glass stopper
  • Glass cap reduces evoporation of alcohol when not in use