Handheld refractometers, AR200
供应商: Reichert
REIC13950000EA 0 CNY
REIC13950000 REIC13952000
Handheld refractometers, AR200
折光仪 手持式折光仪
The AR200 is designed for applications in the food industry and raw material quality assurance across many industries.
  • High accuracy of ±0,0001 nD
  • Results can be downloaded to PC with optional IR communication package
  • Multipoint calibration - Up to five points
Certifications: ICUMSA