Temperature and humidity chart recorders, TH8
供应商: Dickson
DICKTH8P3EA 0 CNY
DICKTH8P3 DICKTH8P5
Temperature and humidity chart recorders, TH8
图表记录器 记录器
These chart recorders with LCD are a fast method for capturing temperature and humidity data. With excellent resolution on Ø 200 mm chart, these models come equipped with an arm holding a pen that swivels upwards, allowing for easy replacement of pen and chart, a digital display and a replaceable probe.
  • User-selectable recording times and temperature ranges
  • Switchable °C/°F and humidity (RH)/dew point (Td) scales
  • User calibration
Delivery information: Supplied with pens, master calibration certificate and back-up AA batteries. Charts need to be ordered separately.