Tubing, silicone, Versilic®
供应商: Saint-Gobain
VERN760210EA 0 CNY
VERN760690-10 VERN760210-10 VERN760510-10 VERN760720 VERN760420 VERN760220 VERN760440 VERN760810-10 VERN760320 VERN760581-10 VERN760580 VERN760800-10 VERN760350-10 VERN761050 VERN761270 VERN760160 VERN761150 VERN760500-10 VERN761170 VERN760650-10 VERN761190 VERN760570-10 VERN760440-10 VERN760800 VERN760820 VERN760230-10 VERN760730 VERN760630 VERN760410 VERN760330 VERN760770 VERN761100 VERN760110 VERN760430 VERN760210 VERN760570 VERN760010 VERN760230 VERN760590 VERN760220-25 VERN760170 VERN760670-10 VERN760490 VERN760070 VERN761080 VERN760520-10 VERN760420-10 VERN760770-10
Tubing, silicone, Versilic®
Silicone, translucent, peroxide cross-linked.
  • Good physical properties such as pliancy, elasticity and flexibility
  • Suitable for use with foodstuffs in accordance with FDA and BgVV
  • Produced in compliance with GMP conditions, ISO 9001 and EN 46002 (CE marking)
For laboratory and medical applications, pharmacies, biotechnology, fermentation and pharmaceutical production. Biologically neutral (USP XXIII class 6).
Temperature resistance: -50 to +200 °C
Shore hardness: 62
Density: 1,16
Certifications: Meets FDA, European and French regulations for food contact