带边试管
供应商: VWR
VWR®
VWRI212-0306EA 380.05 CNY
VWRI212-0306 VWRI212-0022
带边试管
管子 试验/取样管
FIOLAX®,中性玻璃
  • Excellent chemical resistance
  • Relatively resistant to rapid temperature changes and local heating
The economical alternative for a wide range of uses. Thin walled.
Certifications: DIN 12395