Nunc-Immuno™ stick
供应商: Thermo Fisher Scientific
Nunc-Immuno™
NUNC472230EA 0 CNY
NUNC472230
Nunc-Immuno™ stick
管子 免疫管
PP tube and stick, PS paddle. Developed for ELISA and RIA tests in the field.
  • Gives reliable, semi-quantitative results with built-in negative control
  • Hydrophilic MaxiSorp™ paddle surface
  • PP tube for minimal adsorption of assay components
Paddle surface area: 5,2 cm²
External L×W (including lid): 49×12 mm