Sealing film for PCR plates, Axygen® UltraClear
供应商: Corning
Axygen®
AXYGUC-500EA 0 CNY
AXYGUC-500
Sealing film for PCR plates, Axygen® UltraClear
密封件 微孔板封板膜和铝箔
For sealing 96 and 384-well PCR plates.
  • Film clarity allows for optical analysis during Real-Time PCR
  • Unique pressure sensitive adhesive that leaves no tacky residue
  • Functional temperature range −40 to +104 °C