VWR®, β辐射箱
供应商: VWR
VWR®
VWRI113-8830EA 7183.97 CNY
VWRI113-8830
VWR®, β辐射箱
Acrylic, transparent, thickness: 10 mm.
  • With hinged lid
  • Racks for 32 microcentrifuge tubes 0,5/1,5 or 2,0 ml are available
  • Stable and easy to clean
  • 带铰链盖
  • 可容纳32个0.5/1.5或2.0ml微量离心管
  • 稳定且易于清洁
Ideal for storage and transport of test tubes containing radioactive solutions.
订购须知 Box supplied without racks. Racks must be ordered separately.