Micro pipettes, intraMARK, BLAUBRAND®
供应商: Brand
BLAUBRAND®
BRND708709EA 0 CNY
BRND708757_U BRND708709 BRND708707 BRND708718 BRND708728 BRND708757 BRND708745 BRND708733 BRND708722 BRND708744
Micro pipettes, intraMARK, BLAUBRAND®
移液管和移液器 微血管
Glass, with ring mark, disposable.
  • Calibrated to contain 'In', contains the stated volume when filled from one end to a ring mark
  • Colour coded to ISO for clear identification
  • DE-M marking for volumes ≥5 µl
Accuracy ≤±0,25%; Reproducibility ≤0,5% (except for 612-1401: Accuracy ≤±0,30%; Reproducibility ≤0,6%).
Caution: DE-M marking only for 5 µl.