Micro haematocrit capillaries
供应商: Brand
BRND749321EA 0 CNY
BRND749321 BRND749311
Micro haematocrit capillaries
移液管和移液器 微血管
Without calibration mark. For use in haematocrit centrifuges.
  • Heparinised or non-heparinised
  • With colour code
Type A: heparinised with sodium heparinate over internal surface, red colour code, for capillary blood.
Type B: non-heparinised, blue colour code, for use with heparinised venous blood.
Thickness wall: 0,2 mm
Certifications: According to DIN ISO 12772 and BS 4316 - 68.