PCR plates, low profile
供应商: Simport
SIMPT323-96LPNEA 0 CNY
SIMPT323-96LPY SIMPT323-96LPG SIMPT323-96LPR SIMPT323-96LPB SIMPT323-96LPN
PCR plates, low profile
微孔板 PCR板
PP. Thin walled 96-well PCR plates with small tube volume (100 µl).
  • Alphanumeric grid for easy sample identification
  • Flat surface for better sealing
  • Can be cut to desired format
  • Small volume reducing dead space between sample and cover
Thin walled 96-well PCR plates with small tube volume (100 µl).
  • Alphanumeric grid for easy sample identification
  • Flat surface for better sealing
  • Can be cut to desired format
  • Small volume reducing dead space between sample and cover
Packaging: Packed in tamperproof, resealable bags of 10 plates.