VWR® Desiccators, Grease-free
供应商: Avantor
JENC250-068EA 0 CNY
JENC250-068 JENC250-039BOM JENC250-028BOM JENC250-045BOM JENC250-030BOM JENC250-032BOM JENC250-042BOM
VWR® Desiccators, Grease-free
干燥器 玻璃干燥器
Borosilicate glass. Elastomer ring is compressed under vacuum making a dry seal. The PTFE valve is situated centrally on the lid, which in conjunction with the dry seal, enables the lid to be lifted on and off vertically.
  • Choice of models with or without Seal-Cote plastic protection, which prevents fragmentation of the glass
  • Lids lift off vertically, giving nearly double the storage volume
Certifications: BS 3423:1962
Delivery information: Supplied with sealing ring, gauze, and PTFE stopcock. 9 mm O.D. vacuum connection.