Burettes, pellet pattern, lateral stopcock, Schellbach
供应商: Hirschmann Laborgeräte
HIRS3410360EA 0 CNY
HIRS3410360 HIRS3410370 HIRS3410160 HIRS3410170 HIRS3410375 HIRS3410175
Burettes, pellet pattern, lateral stopcock, Schellbach
滴定管
DURAN®, class B.
  • Calibrated to deliver 'Ex'
  • Schellbach stripe, short marks at major graduations, blue graduations
  • Automatic zeroing, without intermediate stopcock
DIN 12 700 T5
Certifications: DIN 12 700 T5
Delivery information: Delivered with 2 L bottle.