Waste disposal bags, Labsak, Jencons
供应商: Jencons
JENC121-024EA 0 CNY
JENC121-024
Waste disposal bags, Labsak, Jencons
废物处理袋
Laminated polypropylene bag for general purpose laboratory and workshop use. Ideal for disposal purposes.
  • Puncture and tear resistant
  • Long term resistance to heat and UV and stabilised for weathering