MethoCult™ M3630 Medium
供应商: STEMCELL Technologies
MethoCult™
STMC03630EA 0 CNY
STMC03630
MethoCult™ M3630 Medium
细胞培养基
Methylcellulose-based medium for mouse pre-B lymphoid progenitor cells.
MethoCult™ M3630 is optimised for the growth and enumeration of mouse pre-B lymphoid progenitor cells in colony-forming unit (CFU) assays of mouse bone marrow and Whitlock-Witte long-term cultures.