2-硝基苯乙腈 98%

供应商: Thermo Scientific
Warning

ACRO172420100EA 0 CNY
ACRO172420500 ACRO172420100
2-硝基苯乙腈 98%
2-硝基苯乙腈
分子式: C₈H₆N₂O₂
分子量: 162,15 g/mol
沸点: 304 °C (1013 hPa)
溶点: 81…85 °C
存储条件: Ambient
MDL编号: MFCD00007183
CAS号: 610-66-2
EINECS: 210-231-0
ADR: 6.1,III

现在订购

产品技术规格

Appearance Light brown to grey-brown Powder
Infrared spectrum Conforms
Melting point 81 to 85 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR