Scintillation Vials, Borosilicate Glass, with White Urea Caps
供应商: Thermo Fisher Scientific
TFBR03-337-5EA 0 CNY
TFBR03-337-5
Scintillation Vials, Borosilicate Glass, with White Urea Caps
小瓶
These scintillation vials, made up of borosilicate glass, with white urea caps fit all common counters.
  • Potassium-free glass provides consistently low activity counts and excellent light transmission
  • Caps fit GPI 22-400 thread finish and are suitable for marking
  • Ext.Ø×H (with cap): 28×61 mm
Packaging: Vials with caps attached are shrink-wrapped in five trays of 100 per case. When caps are separate, vials are shrink-wrapped in five trays of 100 per case, and caps are packed in five polybags of 100 caps each.