Matrix® Safety Face Shield
供应商: MCR Safety
Matrix®
MCRS483000_U1EA 0 CNY
MCRS483000_U1
Matrix® Safety Face Shield
面罩
MCR Safety's headgear and face shields offer superior protection from liquid splashes and flying debris that would otherwise be damaging to the face. An assortment of brackets, face shield sizes, and materials make it a good bet that you can find an option suitable for you. (Headgear only, face shield not included).
  • Head gear for double matrix face shields
  • Ratchets to fit
  • Rugged polycarbonate material
  • Formerly known as Crews® safety glasses
Certifications: This product meets the ANSI/ISEA Z87.1 2020 standard.