VWR® Dylatherm™ Hotplate
供应商: Avantor
VWR® Dylatherm™
VWRU12620-980EA 0 CNY
VWRU12620-980
VWR® Dylatherm™ Hotplate
加热器 平板加热器
The economical Dylatherm™ Hot plate features a large 165 mm diameter cast aluminum top plate that provides an even heating surface capable of heating 1500 ml of water.
  • Built-in support rod holder
  • Bimetallic thermostat offers reliable and repeatable temperature control
Built-in support rod holder with locking knob accepts optional support rod and clamp kit for mounting thermometers and temperature probes. Optional support plate allows hot plate to be mounted to lab frames or ring stands.
Powerful heater reaches maximum temperature in minutes. Indicator light illuminates when unit is on.
Certifications: RoHS certified.
Delivery information: Units include a 1,8 m Euro type plug, 457 mm support rod, thermometer extension clamp, three-prong medium swivel clamp, and hook connector.