Rugged, compact hotplates with an ergonomic design are ideal for educational labs and can heat up to 1000 ml of water.
- Rugged aluminum top plate
- Boils 300 ml of water in 16 minutes
- Built-in support rod holder
Durable 127×127 mm cast aluminum top plate will not chip or crack, and provides a uniform heating surface. The fixed temperature hotplate features an illuminated rocker switch to activate the preset fixed temperature of 375 °C.
Bi-metallic thermostat offers reliable temperature control. Powerful heater reaches maximum temperature in minutes. The built-in support rod holder with locking knob accepts an optional support rod and clamp kit. Optional support plate allows hotplate to be mounted to lab frames or ring stands.
Certifications: TUV listed. RoHS certified.
订购须知 All units include a five-year limited warranty on parts and labour.
Delivery information: Kit includes a 1,83 m cord with a Euro style plug, 457 mm support rod, thermometer extension clamp, three-prong medium swivel clamp, and hook connector.