氢氧化钠 0,9 - 1,1 M

供应商: THERMO FISHER BRAND

Synonyms: 苛性钠

M-Clarity™ quality level = MQ100

TFBRSS266-1EA 0 CNY
TFBRSS266-1
氢氧化钠 0,9 - 1,1 M
氢氧化钠
分子式: NaOH
分子量: 40 g/mol
MDL编号: MFCD00003548
CAS号: 1310-73-2

现在订购

产品技术规格

Appearance (colour) Colorless
Appearance (form) Liquid
Titration (Molarity) 0.9000 - 1.1000 mol/L
UV Absorbance UV-Absorption at 260 nm ≤ 0.500
UV Absorbance UV-Absorption at 270 nm ≤ 0.200
UV Absorbance UV-Absorption at 280 nm ≤ 0.030
UV Absorbance UV-Absorption at 290 nm ≤ 0.010
UV Absorbance UV-Absorption at 500 nm ≤ 0.010
Luminescence Corresponds to requirement
Residue (filter test) No residue
Filtertest (method) Neat
Recommended retest period 84 months

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR