Safety Face Screen
供应商: Honeywell Safety
HONEBD-176B/SE173AEA 0 CNY
HONEBD-176B/SE173A
Safety Face Screen
面罩
Use for safety helmet.
  • Standard : ANSI Z87.1
  • Visible length: 200×390 mm
  • Browguard: Nylon
  • Chinguard: Polycarbonate clear