Sodium formate ≥99%, crystals ACS

供应商: Spectrum Chemical

Synonyms: 蚁酸

SPCMS1285-125GMEA 0 CNY
SPCMS1285-125GM
Sodium formate ≥99%, crystals ACS
甲酸钠

Sodium Formate, Crystal, Reagent, ACS is the sodium salt of formic acid and appears as a deliquescent white powder. It is typically used as a buffering agent for mineral acids, as well as food additive.


As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

分子式: HCO₂Na
分子量: 68,01 g/mol
溶点: 257,31 °C
密度: 1,919 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00013101
CAS号: 141-53-7
EINECS: 205-488-0
REACH: 01-2119486468-21
Merck Index: 13,08694

现在订购

产品技术规格

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR