Nonmetallic Liquid Disposal Cans
供应商: Justrite
JUSU14762EA 0 CNY
JUSU14762
Nonmetallic Liquid Disposal Cans
安全罐
High-density polyethylene cans for containment of corrosives and liquid flammable wastes.
  • High-density polyethylene cans safely contain corrosives or liquid flammable wastes
  • Flame arrester guards against outside ignition
  • Built-in carbon insert provides grounding path
  • Large openings are strategically placed for controlled filling and pouring
  • Fixed-position handle keeps lid open for easier pouring
  • When closed, lid automatically vents at 5 psig
  • FM approved
Large openings are strategically placed for controlled filling/pouring during disposal. Fixed-position handle allows lid to stay open for easier pouring. Closed, the cap automatically vents at 5 psig. Flame arrester guards against outside ignition. Built-in carbon insert provides a grounding path. Plated steel fittings are ideal for use with flammable liquids. Stainless steel fittings are designed for use with corrosives.
Certifications: FM approved.