VWR® 96-well microplates, 500 µl
供应商: VWR
VWR®
VWRIEZ9604-01EA 0 CNY
VWRIEZ9604-01
VWR® 96-well microplates, 500 µl
微孔板 高温超导微孔板
This flat-bottom 96-well microplate fits well with the E-Z 96® magnetic separation device and can be used as a processing plate during DNA and RNA preparation with magnetic bead systems.