J.T.Baker®, Microplate Lid
供应商: Avantor
J.T.Baker®
BAKR94001-0230EA 0 CNY
BAKR94001-0230
J.T.Baker®, Microplate Lid
盖子 Microplate Lids
Polystyrene lid suitable for use with J.T.Baker® 384 and 96 SW.
  • Material: Polystyrene
  • Dimensions (outside): 129,2×86,0×9,0 mm