No-touch hand cleaner dispensers, ASEPTI-CLEANSE®
供应商: Veltek
ASEPTI-CLEANSE®
VELTDEC-301EA 6519.95 CNY
VELTDEC-301
No-touch hand cleaner dispensers, ASEPTI-CLEANSE®
Soap Dispensers and Lotion Soap Dispensers
ASEPTI-CLEANSE® is a hands-free dispensing system with the most advanced infrared sensor which can be set to dispense approximately 1, 3 or 5 mL of solution.
  • Dispenses a pre-measured dose to the hand without user contact
  • Photo-eye operated
  • Water resistant design
  • Operates on 4 D-Cell Batteries or 110 V direct connections
ASEPTI-CLEANSE® has been designed by VAI as the most advanced Infrared Sensor Dispensing System available in the pharmaceutical and biotech Asepti-Cleanseindustry. The dispensing unit is designed to
meet the requirements of cGMP clean room operations.
Excellent choice for gowning rooms and aseptic manufacturing areas.