Positive Displacement Utility Drill Pumps, HDPE
MFLS75501-31EA 0 CNY
MFLS75501-31
Positive Displacement Utility Drill Pumps, HDPE
Diaphragm Pumps
Connects to a standard electric drill.
  • Inlet and outlet connections accept 3/4" garden hoses
  • Self-priming up to 6 ft (1.8 m)
  • Pumps up to 156 GPH (at 2500 rpm)
This Positive Displacement Utility Drill Pump is constructed of HDPE for applications requiring enhanced chemical resistance.