Great Plains Economy Fuel and Water Meters
供应商: Great Plains Industries
MFLB32486-02EA 0 CNY
MFLB32486-02 MFLB32485-99 MFLB32486-01 MFLB32486-00
Great Plains Economy Fuel and Water Meters
流量计
Small, powerful, and durable.
  • Battery powered 4 digit display
  • Factory calibrated to water or fuel
  • Gallon or litre measurement units
  • Batch and cumulative totals
  • Ready to install
Small, powerful, microprocessor based totalizers powered by AAA batteries have large four-digit displays and mount at the nozzle for ultimate convenience. They are suited for flow ranges from 3 to 30 GPM. Nylon® models, rated to 150 PSIG, are factory calibrated for water. Aluminum models, rated to 300 PSIG are factory calibrated for fuels. Accuracy is ±5%. All mount with 1 inch female NPT fittings.