Advanced Variable-Speed DC Motor Drives
MFLB70100-00EA 0 CNY
MFLB70100-00
Advanced Variable-Speed DC Motor Drives
Motors
Vary the speed of your DC motor with these controllers.
  • Easily control the speed of your DC motor
Advanced controller features three control modes-manual, remote, and proportional remote. It accepts 0 to 10 VDC or 4 to 20 mA analog input and adjusts speed of 1/4- to 2-hp motors without additional resistors or other modifications. The NEMA 4/12 (IP 52/56) housing allows safe operation in damp, dusty work areas.
Delivery information: Heat sink for thermal overload protection.