FMI Series Q Pump Head Adapter for 56C Mount Motors
供应商: FMI
MFLA07117-85EA 0 CNY
MFLA07117-85
FMI Series Q Pump Head Adapter for 56C Mount Motors
Piston Pumps
Configurable to your requirements.
  • Easily adapt and mount your Series Q pump head
Operate the Series Q pump head (sold on the opposite page) with Masterflex L/S™ drives (sold on pages 787-806) and NEMA Type 56C-face motors (sold on pages 897-899) using these adapter kits.