VWR®, Digital 4 Plate Incubated Shaker
供应商: VWR
VWR®
VWRA444-1223EA 0 CNY
VWRA444-1223
VWR®, Digital 4 Plate Incubated Shaker
振荡器 微孔板振荡器
This device has been deisgned for performance and durability, with the objective of delivering perfectionism. One of the key feature is the maintenance free brushless DC motor, which performs efficiently even under high loads for thorough mixing.
  • Temperature setting range from Ambient +3 to 70 °C
  • Adjustable speed ranging from 200 to 1200 min⁻¹
  • Microprocessor controlled Thermo Shaker with variable speed and time setting and also with last run memory function
  • Long timer setting up to 99 hours and 59 minutes
  • Critical parameters like speed, time as well as set and actual temperature are displayed digitally
  • Rubber O-ring provides firm grip for securing the microplate
  • Programmable pulse mode (30 sec to 99 sec) for vigorous shaking in clockwise and counter-clockwise direction
  • Easy to clean and maintain the equipment
  • With in-built counter balance
Unit is compatible with all skirted microplates and has a maximum capacity of 4 plates.