甲醇 ≥99.5% (通过GC) NF, Macron Fine Chemicals™

供应商: Avantor Performance Materials
Danger

Synonyms: 木精

Management of Change (MOC) category = RL

MACR8888-REA 0 CNY
MACR8888-R
甲醇 ≥99.5% (通过GC) NF, Macron Fine Chemicals™
甲醇

Tested to NF Specifications (for Upsher-Smith)


No Class 1 or Class 2 Residual Solvents are present in this product Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0,5%.

分子式: CH₃OH
分子量: 32,04 g/mol
MDL编号: MFCD00004595
CAS号: 67-56-1
UN号: 1230
ADR: 3,II

现在订购

产品技术规格

For Laboratory,Research,or Manufacturing Use
NF - Assay (CH₃OH) (by GC) ≥99.5 %
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Acetone, Aldehydes (as (CH₃)₂CO) ≤0.003 %
NF - Acidity Passes Test
NF - Alkalinity (as NH₃) ≤3 ppm
NF - Nonvolatile Residue ≤0.001 %
NF - Readily Carbonizable Substances Passes Test
NF - Readily Oxidizable Substances Passes Test
NF - Water (H₂O) ≤0.1 %
No Class 1 or Class 2 Residual Solvents are present in this product Only
Class 3 Solvents are likely to be present. Residual concentration of all
Class 3 Solvents is <0.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR