氯化钠 99.0-100.5% (基于干燥的物质), crystals USP, FCC, J.T.Baker®

供应商: Avantor
Warning

BAKR3302-25EA 0 CNY
BAKR3302-25 BAKR3302-QM
氯化钠 99.0-100.5% (基于干燥的物质), crystals USP, FCC, J.T.Baker®
氯化钠
分子式: NaCl
分子量: 58,44 g/mol
溶点: 801 °C
密度: 2,16 g/ml (25 °C)
MDL编号: MFCD00003477
CAS号: 7647-14-5

现在订购

产品技术规格

GMP Manufactured Product
Meets F.C.C. Requirements
Meets U.S.P Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Acidity or Alkalinity Passes Test
USP - Appearance of Solution Passes Test
USP - Aluminum (Al) ≤0.2 ppm
USP - Arsenic (As) ≤1 ppm
USP - Barium (Ba) Passes Test
USP - Bromide (Br) ≤100 ppm
USP - Ferrocyanide Passes Test
USP - Iodides (I) Passes Test
USP - Iron (Fe) ≤2 ppm
USP - Loss on Drying at 105°C ≤0.5 %
USP - Magnesium and Alkaline Earth Metals (as Ca) ≤100 ppm
USP - Nitrite (NO₂) (Absorbance Value) ≤0.01
USP - Phosphate (PO₄) ≤25 ppm
USP - Potassium (K) ≤500 ppm
USP - Sulfate (SO₄) ≤200 ppm
USP - Assay (NaCl) (dried basis) 99.0 - 100.5 %
FCC - Assay (as NaCl) (FCC) (ignited basis) 99.0 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Arsenic (As) ≤1 ppm
FCC - Calcium and Magnesium ≤0.35 %
FCC - Heavy Metals (as Pb) ≤2 ppm
FCC - Loss on Drying at 105°C ≤0.5 %
Solution Test Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Must be subjected to further processing during the preparation of
injectable dosage forms
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR