VWR®, Scoop Type Spatula
供应商: VWR
VWR®
VWRU470149-438EA 0 CNY
VWRU470149-438
VWR®, Scoop Type Spatula
样品勺
This scoop-type spatula is versatile and durable.
  • Semicircular cross section
  • Stainless steel construction
  • Spatula dimensions: ¹/₂×7¹/₂"
It also has one rounded end and one pointed end.