2,2,4-三甲基戊烷 ≥99.0% (通过GC,校正水含量), PHOTREX® 用于分光光度法, J.T.Baker®

Danger

Synonyms: 异辛烷

Management of Change (MOC) category = NI

BAKR9479-03EA 0 CNY
BAKR9479-03
2,2,4-三甲基戊烷 ≥99.0% (通过GC,校正水含量), PHOTREX® 用于分光光度法, J.T.Baker®
2,2,4-三甲基戊烷
分子式: (CH₃)₂CHCH₂C(CH₃)₃
分子量: 114,23 g/mol
沸点: 99,3 °C
密度: 0,690 g/cm³ (25 °C)
闪点: –12,00 °C
MDL编号: MFCD00008943
CAS号: 540-84-1
UN号: 1262
ADR: 3,II

现在订购

产品技术规格

For Laboratory,Research,or Manufacturing Use
ACS - Assay ((CH₃)₃CCH₂CH(CH₃)₂) (by GC, corrected for water) ≥99.0 %
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.0005 %
ACS - Sulfur Compounds (as S) ≤0.005 %
ACS - Water (by KF, coulometric) ≤0.05 %
ACS - Water-Soluble Titrable Acid, meq/g ≤0.0003
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-250 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm ≤0.04
Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.20
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤1.00
Product Information (not specifications):
Windows of Infrared Transmittance (0.1-mm path, 50-100% T), µm: 2.5 - 3.3; 3.6 - 6.6; 7.5 - 7.7; 8.7 - 10.2; 10..3 - 15.0
Boiling Point (typical)................. 99.3 °C.
Density (g/mL) at 25 °C (typical)........ 0.690

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR