G418 disulphate

供应商: INVITROGEN
Warning

Synonyms: 抗生物質G418

INVT10131027EA 0 CNY
INVT10131027
G418 disulphate
G 418
分子式: C₂₀H₄₄N₄O₁₈S₂
分子量: 692,72 g/mol
存储条件: Ambient
MDL编号: MFCD00058314
CAS号: 108321-42-2

现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR