Solvent ASTM D235 Type 1 适用于通过ASTM方法测试石油产品
供应商: VWR Chemicals
|
Danger
|
VWRC30694HCEA
0
CNY
VWRC30694HC
VWRC306946X
Solvent ASTM D235 Type 1 适用于通过ASTM方法测试石油产品
Solvent ASTM D235 Type 1
White spirit (BSS 245) Type A 1976
沸点:
135…220 °C (1013 hPa) 溶点: –15 °C 密度: 0,79 g/cm³ (20 °C) 闪点: 30 °C (closed cup) |
UN号:
1300 ADR: 3,III |
产品技术规格
Density (15/4) | 0.770 - 0.790 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...