VWR®, Low Profile Magnetic Stirrer
供应商: VWR
VWR®
VWRA10153-690EA 0 CNY
VWRA10153-690
VWR®, Low Profile Magnetic Stirrer
搅拌器 磁力搅拌器
The VWR® low profile magnetic stirrer is a laboratory staple.
  • Compact and sleek design
  • Variable speed: 15 to 1500 min⁻¹
  • Designed with a high level of waterproofing protection
The small footprint and sleek design save valuable bench space, while the unit provides excellent stirring capability for many routine applications.
订购须知 Supplied without magnetic stirring bars.