Vacuum pump, EZ-Stream
供应商: Merck
MILFEZSTREAM1EA 0 CNY
MILFEZSTREAM1
Vacuum pump, EZ-Stream
真空泵
This laboratory filtration pump is ideal to transfer filtered liquids directly to waste.
  • Flow rate: 3,8 to 4,0 L/min
Used to transfer beer, dairy, bottled water, food and beverage, pharmaceutical products analysis, soft drink, cider, sport drink and wine.
Delivery information: Includes pump, multilingual user guide and power supply.